Assay Details
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | UGCUAUGCCAACAUAUUGCCAUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-31-3p | MIMAT0004634 |
Rat | rno-miR-31a-3p | MIMAT0017102 |
Cricetulus griseus | cgr-miR-31-3p | MIMAT0023903 |
Dasypus novemcinctus | dno-miR-31-3p | MIMAT0047568 |
Guinea pig | cpo-miR-31-3p | MIMAT0046915 |
Rabbit | ocu-miR-31-3p | MIMAT0048162 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Rabbit
