Assay Details
Mature miRNA Sequence: | AACUGGCCUACAAAGUCCCAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chrysemys picta | cpi-miR-193a-3p | MIMAT0037810 |
| Gorilla gorilla | ggo-miR-193a | MIMAT0024174 |
| Monodelphis domestica | mdo-miR-193a-3p | MIMAT0004123 |
Stem-loop Details
Chrysemys picta
Gorilla gorilla
Monodelphis domestica
More Information
Related Products
miRBase Alias:
Human: hsa-miR-193a (v9.2)
Mouse: mmu-miR-193 (v9.2)
Rat: rno-miR-193 (v9.2)
Monodelphis domestica: mdo-miR-193 (v19)
Mouse: mmu-miR-193 (v9.2)
Rat: rno-miR-193 (v9.2)
Monodelphis domestica: mdo-miR-193 (v19)

Genomic Map