Assay Details
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | UAGCAGCACAGAAAUAUUGGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-195-5p | MIMAT0000461 | |
| Mouse | mmu-miR-195a-5p | MIMAT0000225 | |
| Rat | rno-miR-195-5p | MIMAT0000870 | |
| Gorilla gorilla | ggo-miR-195 | MIMAT0002316 | |
| Horse | eca-miR-195 | MIMAT0013027 | |
| Pan paniscus | ppa-miR-195 | MIMAT0002317 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-195 | MIMAT0008060 | |
| Pig | ssc-miR-195 | MIMAT0013928 | |
| Pongo pygmaeus | ppy-miR-195 | MIMAT0015790 | |
| Rhesus monkey | mml-miR-195-5p | MIMAT0006228 | |
Stem-loop Details
Human
Mouse
Rat
Gorilla gorilla
Horse
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-195 (v17)
Mouse: mmu-miR-195 (v17)
Mouse: mmu-miR-195-5p (v18)
Rat: rno-miR-195 (v18)
Macaca mulatta: mml-miR-195 (v19)
Mouse: mmu-miR-195 (v17)
Mouse: mmu-miR-195-5p (v18)
Rat: rno-miR-195 (v18)
Macaca mulatta: mml-miR-195 (v19)

Genomic Map