Assay Details
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | UAGCAGCACAGAAAUAUUGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-195-5p | MIMAT0000461 |
| Mouse | mmu-miR-195a-5p | MIMAT0000225 |
| Rat | rno-miR-195-5p | MIMAT0000870 |
| Gorilla gorilla | ggo-miR-195 | MIMAT0002316 |
| Horse | eca-miR-195 | MIMAT0013027 |
| Pan paniscus | ppa-miR-195 | MIMAT0002317 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-195 | MIMAT0008060 |
| Pig | ssc-miR-195 | MIMAT0013928 |
| Pongo pygmaeus | ppy-miR-195 | MIMAT0015790 |
| Rhesus monkey | mml-miR-195-5p | MIMAT0006228 |
Stem-loop Details
Human
Mouse
Rat
Gorilla gorilla
Horse
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-195 (v17)
Mouse: mmu-miR-195 (v17)
Mouse: mmu-miR-195-5p (v18)
Rat: rno-miR-195 (v18)
Macaca mulatta: mml-miR-195 (v19)
Mouse: mmu-miR-195 (v17)
Mouse: mmu-miR-195-5p (v18)
Rat: rno-miR-195 (v18)
Macaca mulatta: mml-miR-195 (v19)

Genomic Map