Assay Details
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | UAGCAGCACAGAAAUAUUGGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-195-5p | MIMAT0000461 |
Mouse | mmu-miR-195a-5p | MIMAT0000225 |
Rat | rno-miR-195-5p | MIMAT0000870 |
Gorilla gorilla | ggo-miR-195 | MIMAT0002316 |
Horse | eca-miR-195 | MIMAT0013027 |
Pan paniscus | ppa-miR-195 | MIMAT0002317 |
View More Rows | ||
Pan troglodytes | ptr-miR-195 | MIMAT0008060 |
Pig | ssc-miR-195 | MIMAT0013928 |
Pongo pygmaeus | ppy-miR-195 | MIMAT0015790 |
Rhesus monkey | mml-miR-195-5p | MIMAT0006228 |
Stem-loop Details
Human
Mouse
Rat
Gorilla gorilla
Horse
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
