Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AUCACACAAAGGCAACUUUUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-377-3p | MIMAT0000730 | |
| Mouse | mmu-miR-377-3p | MIMAT0000741 | |
| Bovine | bta-miR-377 | MIMAT0009304 | |
| Callithrix jacchus | cja-miR-377 | MIMAT0039379 | |
| Guinea pig | cpo-miR-377-3p | MIMAT0047218 | |
| Horse | eca-miR-377 | MIMAT0013146 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-377 | MIMAT0008129 | |
| Pongo pygmaeus | ppy-miR-377 | MIMAT0015867 | |
| Pteropus alecto | pal-miR-377-3p | MIMAT0040413 | |
| Rabbit | ocu-miR-377-3p | MIMAT0048420 | |
| Rhesus monkey | mml-miR-377-3p | MIMAT0006306 | |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-377 (v17)
Mouse: mmu-miR-377 (v17)
Macaca mulatta: mml-miR-377 (v19)
Ovis aries: oar-miR-377-3p (v19)
Mouse: mmu-miR-377 (v17)
Macaca mulatta: mml-miR-377 (v19)
Ovis aries: oar-miR-377-3p (v19)

Genomic Map