Assay Details
Amplicon Length: | 54 |
Chromosome Location: | Chr. 12 - 109740713 - 109740713 [+] on 109740713 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-377-5p | MIMAT0017079 |
| Mouse | mmu-miR-377-3p | MIMAT0000741 |
Stem-loop Details
Mouse

Genomic Map