Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 14 - 101062245 - 101062245 [+] on 101062245 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-377-5p | MIMAT0004689 |
| Human | hsa-miR-377-3p | MIMAT0000730 |
Stem-loop Details
Human

Genomic Map