Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AUCACACAAAGGCAACUUUUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-377-3p | MIMAT0000730 |
| Mouse | mmu-miR-377-3p | MIMAT0000741 |
| Bovine | bta-miR-377 | MIMAT0009304 |
| Callithrix jacchus | cja-miR-377 | MIMAT0039379 |
| Guinea pig | cpo-miR-377-3p | MIMAT0047218 |
| Horse | eca-miR-377 | MIMAT0013146 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-377 | MIMAT0008129 |
| Pongo pygmaeus | ppy-miR-377 | MIMAT0015867 |
| Pteropus alecto | pal-miR-377-3p | MIMAT0040413 |
| Rabbit | ocu-miR-377-3p | MIMAT0048420 |
| Rhesus monkey | mml-miR-377-3p | MIMAT0006306 |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map