More Information
Updated Mapping Information
miRBase Alias:
Human: hsa-miR-422a (v9.2)
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-422a |
|---|---|
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGGACUUAGGGUCAGAAGGCC |
| Product Type: | TaqMan™ MicroRNA Assay |