Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CAAAGUGCUGUUCGUGCAGGUAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-93-5p | MIMAT0000093 | |
| Mouse | mmu-miR-93-5p | MIMAT0000540 | |
| Rat | rno-miR-93-5p | MIMAT0000817 | |
| Alligator mississippiensis | ami-miR-93-5p | MIMAT0038104 | |
| Callithrix jacchus | cja-miR-93 | MIMAT0039390 | |
| Cricetulus griseus | cgr-miR-93-5p | MIMAT0024013 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-93-5p | MIMAT0047586 | |
| Daubentonia madagascariensis | dma-miR-93 | MIMAT0049285 | |
| Dog | cfa-miR-93 | MIMAT0006696 | |
| Goat | chi-miR-93-5p | MIMAT0036305 | |
| Guinea pig | cpo-miR-93-5p | MIMAT0046933 | |
| Horse | eca-miR-93 | MIMAT0013060 | |
| Nomascus leucogenys | nle-miR-93 | MIMAT0049351 | |
| Otolemur garnettii | oga-miR-93 | MIMAT0049702 | |
| Papio hamadryas | pha-miR-93 | MIMAT0049552 | |
| Pteropus alecto | pal-miR-93-5p | MIMAT0039937 | |
| Rabbit | ocu-miR-93-5p | MIMAT0048176 | |
| Tupaia chinensis | tch-miR-93-5p | MIMAT0036617 | |
| Xenopus laevis | xla-miR-93-5p | MIMAT0046613 | |
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Callithrix jacchus
Cricetulus griseus
Dasypus novemcinctus
Daubentonia madagascariensis
Dog
Goat
Guinea pig
Horse
Nomascus leucogenys
Otolemur garnettii
Papio hamadryas
Pteropus alecto
Rabbit
Tupaia chinensis
Xenopus laevis
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-93 (v17)
Mouse: mmu-miR-93 (v17)
Rat: rno-miR-93 (v18)
Mouse: mmu-miR-93 (v17)
Rat: rno-miR-93 (v18)

Genomic Map