Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CAAAGUGCUGUUCGUGCAGGUAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-93-5p | MIMAT0000093 |
Mouse | mmu-miR-93-5p | MIMAT0000540 |
Rat | rno-miR-93-5p | MIMAT0000817 |
Alligator mississippiensis | ami-miR-93-5p | MIMAT0038104 |
Callithrix jacchus | cja-miR-93 | MIMAT0039390 |
Cricetulus griseus | cgr-miR-93-5p | MIMAT0024013 |
View More Rows | ||
Dasypus novemcinctus | dno-miR-93-5p | MIMAT0047586 |
Daubentonia madagascariensis | dma-miR-93 | MIMAT0049285 |
Dog | cfa-miR-93 | MIMAT0006696 |
Goat | chi-miR-93-5p | MIMAT0036305 |
Guinea pig | cpo-miR-93-5p | MIMAT0046933 |
Horse | eca-miR-93 | MIMAT0013060 |
Nomascus leucogenys | nle-miR-93 | MIMAT0049351 |
Otolemur garnettii | oga-miR-93 | MIMAT0049702 |
Papio hamadryas | pha-miR-93 | MIMAT0049552 |
Pteropus alecto | pal-miR-93-5p | MIMAT0039937 |
Rabbit | ocu-miR-93-5p | MIMAT0048176 |
Tupaia chinensis | tch-miR-93-5p | MIMAT0036617 |
Xenopus laevis | xla-miR-93-5p | MIMAT0046613 |
Stem-loop Details
Human
Stem-loop ID | hsa-mir-93 |
Stem-loop Accession # | MI0000095 |
Stem-loop Sequence | CUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCCAACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCGG |
Chromosome Location | Chr. 7 - 100093768 - 100093847 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000093
miRBase ID: hsa-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: Chr. 7 - 100093768 - 100093847 [-] on Build GRCh38 |
Stem-loop ID | mmu-mir-93 |
Stem-loop Accession # | MI0000581 |
Stem-loop Sequence | AGUCAUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUAAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGGACA |
Chromosome Location | Chr. 5 - 138165523 - 138165610 [-] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000540
miRBase ID: mmu-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: Chr. 5 - 138165523 - 138165610 [-] on Build GRCm38 |
Stem-loop ID | rno-mir-93 |
Stem-loop Accession # | MI0000880 |
Stem-loop Sequence | AGUCAUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGCAUUGCCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGGACA |
Chromosome Location | Chr. 12 - 19307543 - 19307629 [-] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000817
miRBase ID: rno-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: Chr. 12 - 19307543 - 19307629 [-] on Build Rnor_6.0 |
Stem-loop ID | ami-mir-93 |
Stem-loop Accession # | MI0029705 |
Stem-loop Sequence | CUUGAGCUGCGGUGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGUGCAGCCAUCUACUGCCGGGCAGCACUUCCCGAGCCCCCGCAGCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038104
miRBase ID: ami-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | cja-mir-93 |
Stem-loop Accession # | MI0031909 |
Stem-loop Sequence | GUCCUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCCAACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCGGGAC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039390
miRBase ID: cja-miR-93 Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | cgr-mir-93 |
Stem-loop Accession # | MI0020562 |
Stem-loop Sequence | AUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUAAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0024013
miRBase ID: cgr-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | dno-mir-93 |
Stem-loop Accession # | MI0038950 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0047586
miRBase ID: dno-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | dma-mir-93 |
Stem-loop Accession # | MI0040016 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049285
miRBase ID: dma-miR-93 Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | cfa-mir-93 |
Stem-loop Accession # | MI0008110 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006696
miRBase ID: cfa-miR-93 Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | chi-mir-93 |
Stem-loop Accession # | MI0030841 |
Stem-loop Sequence | CUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGAUCACCUGACCUACUGCUGAGCCAGCACUUCCCGAGCCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036305
miRBase ID: chi-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | cpo-mir-93 |
Stem-loop Accession # | MI0038606 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0046933
miRBase ID: cpo-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | eca-mir-93 |
Stem-loop Accession # | MI0012804 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013060
miRBase ID: eca-miR-93 Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | nle-mir-93 |
Stem-loop Accession # | MI0040088 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCCGACCUACUGCUGAGCUAGCACUUCCCGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049351
miRBase ID: nle-miR-93 Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | oga-mir-93 |
Stem-loop Accession # | MI0040486 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049702
miRBase ID: oga-miR-93 Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | pha-mir-93 |
Stem-loop Accession # | MI0040320 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUAACCGACCUACUGCUGAGCUAGCACUUCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049552
miRBase ID: pha-miR-93 Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | pal-mir-93 |
Stem-loop Accession # | MI0032409 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039937
miRBase ID: pal-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | ocu-mir-93 |
Stem-loop Accession # | MI0039261 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0048176
miRBase ID: ocu-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | tch-mir-93 |
Stem-loop Accession # | MI0031290 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036617
miRBase ID: tch-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |
Stem-loop ID | xla-mir-93 |
Stem-loop Accession # | MI0038341 |
Stem-loop Sequence | CAAAGUGCUGUUCGUGCAGGUAGCUUUAUAACAGACCUACUGCAUGAGCAGCACUUCCUGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0046613
miRBase ID: xla-miR-93-5p Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG Chromosome Location: NA |

More Information
Related Products

































