Assay Details
Gene Family ID: | MIPF0000641, mir-591 |
Mature miRNA Sequence: | AGACCAUGGGUUCUCAUUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-591 | MIMAT0003259 |
| Pan troglodytes | ptr-miR-591 | MIMAT0008262 |
| Pongo pygmaeus | ppy-miR-591 | MIMAT0016034 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map