Assay Details
Gene Family ID: | MIPF0000672, mir-617 |
Mature miRNA Sequence: | AGACUUCCCAUUUGAAGGUGGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-617 | MIMAT0003286 |
| Pan troglodytes | ptr-miR-617 | MIMAT0008279 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map