More Information
Updated Mapping Information

Related Products
miRBase Alias:
Mouse: mmu-miR-699 (v12)

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-699 |
---|---|
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGGCAGUGCGACCUGGCUCG |
Product Type: | TaqMan™ MicroRNA Assay |