Assay Details
Gene Family ID: | MIPF0000099, mir-136 |
Mature miRNA Sequence: | CAUCAUCGUCUCAAAUGAGUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-136-3p | MIMAT0004606 | |
| Rat | rno-miR-136-3p | MIMAT0004733 | |
| Dasypus novemcinctus | dno-miR-136-3p | MIMAT0047645 | |
| Guinea pig | cpo-miR-136-3p | MIMAT0046998 | |
| Microcebus murinus | mmr-miR-136 | MIMAT0049156 | |
| Otolemur garnettii | oga-miR-136 | MIMAT0049677 | |
| View More Rows | |||
| Pig | ssc-miR-136-3p | MIMAT0037320 | |
| Pteropus alecto | pal-miR-136-3p | MIMAT0040002 | |
| Rabbit | ocu-miR-136-3p | MIMAT0048233 | |
Stem-loop Details
Human
Rat
Dasypus novemcinctus
Guinea pig
Microcebus murinus
Otolemur garnettii
Pig
Pteropus alecto
Rabbit
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool B v3.0Megaplex™ RT Primers, Rodent Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Rodent MicroRNA B Cards v3.0TaqMan® Human MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Human: hsa-miR-136* (v17)
Rat: rno-miR-136* (v18)
Rat: rno-miR-136* (v18)

Genomic Map