Assay Details
Gene Family ID: | MIPF0000099, mir-136 |
Mature miRNA Sequence: | CAUCAUCGUCUCAAAUGAGUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-136-3p | MIMAT0004606 |
| Rat | rno-miR-136-3p | MIMAT0004733 |
| Dasypus novemcinctus | dno-miR-136-3p | MIMAT0047645 |
| Guinea pig | cpo-miR-136-3p | MIMAT0046998 |
| Microcebus murinus | mmr-miR-136 | MIMAT0049156 |
| Otolemur garnettii | oga-miR-136 | MIMAT0049677 |
| View More Rows | ||
| Pig | ssc-miR-136-3p | MIMAT0037320 |
| Pteropus alecto | pal-miR-136-3p | MIMAT0040002 |
| Rabbit | ocu-miR-136-3p | MIMAT0048233 |
Stem-loop Details
Human
Rat
Dasypus novemcinctus
Guinea pig
Microcebus murinus
Otolemur garnettii
Pig
Pteropus alecto
Rabbit

Genomic Map