Assay Details
Gene Family ID: | MIPF0000074, mir-105 |
Mature miRNA Sequence: | UCAAAUGCUCAGACUCCUGUGGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-105-5p | MIMAT0000102 |
Bovine | bta-miR-105a | MIMAT0009217 |
Horse | eca-miR-105 | MIMAT0013193 |
Pongo pygmaeus | ppy-miR-105 | MIMAT0002450 |
Rhesus monkey | mml-miR-105-5p | MIMAT0002454 |
Stem-loop Details
Human
Bovine
Horse
Pongo pygmaeus
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-105 (v17)
Macaca mulatta: mml-miR-105 (v19)
Macaca mulatta: mml-miR-105 (v19)
