Assay Details
Gene Family ID: | MIPF0000074, mir-105 |
Mature miRNA Sequence: | ACGGAUGUUUGAGCAUGUGCUA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-105-3p | MIMAT0004516 |
Rhesus monkey | mml-miR-105-3p | MIMAT0026583 |
Stem-loop Details
Human
Rhesus monkey
