Assay Details
Gene Family ID: | MIPF0000490, mir-939 |
Mature miRNA Sequence: | UGGGGAGCUGAGGCUCUGGGGGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-939-5p | MIMAT0004982 |
| Pan troglodytes | ptr-miR-939 | MIMAT0008346 |
| Rhesus monkey | mml-miR-939 | MIMAT0006540 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey

Genomic Map