Assay Details
Gene Family ID: | MIPF0000420, mir-891 |
Mature miRNA Sequence: | UGCAACUUACCUGAGUCAUUGA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-891b | MIMAT0004913 |
Pan troglodytes | ptr-miR-891b | MIMAT0008334 |
Pongo pygmaeus | ppy-miR-891b | MIMAT0016127 |
Rhesus monkey | mml-miR-891a | MIMAT0006530 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
