Assay Details
Gene Family ID: | MIPF0000199, mir-331 |
Mature miRNA Sequence: | CUAGGUAUGGUCCCAGGGAUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-331-5p | MIMAT0004700 | |
| Mouse | mmu-miR-331-5p | MIMAT0004643 | |
| Dasypus novemcinctus | dno-miR-331-5p | MIMAT0047813 | |
| Guinea pig | cpo-miR-331-5p | MIMAT0047168 | |
| Pongo pygmaeus | ppy-miR-331-5p | MIMAT0015835 | |
| Rabbit | ocu-miR-331-5p | MIMAT0048380 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-331-5p | MIMAT0006272 | |
Stem-loop Details
Human
Mouse
Dasypus novemcinctus
Guinea pig
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products

Genomic Map