Assay Details
Gene Family ID: | MIPF0000199, mir-331 |
Mature miRNA Sequence: | CUAGGUAUGGUCCCAGGGAUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-331-5p | MIMAT0004700 |
| Mouse | mmu-miR-331-5p | MIMAT0004643 |
| Dasypus novemcinctus | dno-miR-331-5p | MIMAT0047813 |
| Guinea pig | cpo-miR-331-5p | MIMAT0047168 |
| Pongo pygmaeus | ppy-miR-331-5p | MIMAT0015835 |
| Rabbit | ocu-miR-331-5p | MIMAT0048380 |
| View More Rows | ||
| Rhesus monkey | mml-miR-331-5p | MIMAT0006272 |
Stem-loop Details
Human
Mouse
Dasypus novemcinctus
Guinea pig
Pongo pygmaeus
Rabbit
Rhesus monkey

Genomic Map