Assay Details
Amplicon Length: | 94 |
Chromosome Location: | Chr. 10 - 93964069 - 93964069 [+] on 93964069 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-331-5p | MIMAT0004643 |
| Mouse | mmu-miR-331-3p | MIMAT0000571 |
Stem-loop Details
Mouse

Genomic Map