Assay Details
Amplicon Length: | 115 |
Chromosome Location: | Chr. 12 - 95308220 - 95308220 [-] on 95308220 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-331-3p | MIMAT0000760 |
| Human | hsa-miR-331-5p | MIMAT0004700 |
Stem-loop Details
Human

Genomic Map