Assay Details
Gene Family ID: | MIPF0000082, mir-193 |
Mature miRNA Sequence: | AACUGGCCUACAAAGUCCCAGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-193a-3p | MIMAT0000459 | |
Mouse | mmu-miR-193a-3p | MIMAT0000223 | |
Rat | rno-miR-193a-3p | MIMAT0000868 | |
Astatotilapia burtoni | abu-miR-193-3p | MIMAT0042161 | |
Bovine | bta-miR-193a-3p | MIMAT0003795 | |
Chicken | gga-miR-193a-3p | MIMAT0007740 | |
View More Rows | |||
Cricetulus griseus | cgr-miR-193a | MIMAT0041152 | |
Cyprinus carpio | ccr-miR-193a | MIMAT0026251 | |
Dasypus novemcinctus | dno-miR-193a-3p | MIMAT0047722 | |
Eptesicus fuscus | efu-miR-193a | MIMAT0035142 | |
Fugu rubripes | fru-miR-193 | MIMAT0002969 | |
Guinea pig | cpo-miR-193a-3p | MIMAT0047071 | |
Horse | eca-miR-193a-3p | MIMAT0013026 | |
Metriaclima zebra | mze-miR-193 | MIMAT0042391 | |
Neolamprologus brichardi | nbr-miR-193 | MIMAT0042573 | |
Ophiophagus hannah | oha-miR-193-3p | MIMAT0036780 | |
Oreochromis niloticus | oni-miR-193 | MIMAT0042763 | |
Otolemur garnettii | oga-miR-193a | MIMAT0049716 | |
Pan paniscus | ppa-miR-193a | MIMAT0049094 | |
Pan troglodytes | ptr-miR-193a | MIMAT0008058 | |
Papio hamadryas | pha-miR-193a | MIMAT0049518 | |
Pig | ssc-miR-193a-3p | MIMAT0013895 | |
Pongo pygmaeus | ppy-miR-193a-3p | MIMAT0015788 | |
Pteropus alecto | pal-miR-193a-3p | MIMAT0039932 | |
Python bivittatus | pbv-miR-193a-3p | MIMAT0038990 | |
Rabbit | ocu-miR-193a-3p | MIMAT0048304 | |
Rhesus monkey | mml-miR-193a-3p | MIMAT0006226 | |
Tetraodon nigroviridis | tni-miR-193 | MIMAT0002970 | |
Tupaia chinensis | tch-miR-193a-3p | MIMAT0036584 | |
Zebrafish | dre-miR-193a-3p | MIMAT0001856 |
Stem-loop Details
Human
Stem-loop ID | hsa-mir-193a |
Stem-loop Accession # | MI0000487 |
Stem-loop Sequence | CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUUCUCGGCCCCCG |
Chromosome Location | Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000459
miRBase ID: hsa-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38 |
Stem-loop ID | mmu-mir-193a |
Stem-loop Accession # | MI0000235 |
Stem-loop Sequence | GAGAGCUGGGUCUUUGCGGGCAAGAUGAGAGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCCUC |
Chromosome Location | Chr. 11 - 79711969 - 79712034 [+] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000223
miRBase ID: mmu-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: Chr. 11 - 79711969 - 79712034 [+] on Build GRCm38 |
Stem-loop ID | rno-mir-193a |
Stem-loop Accession # | MI0000936 |
Stem-loop Sequence | GCGGACGGGAGCUGAGAGCUGGGUCUUUGCGGGCAAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCUCCC |
Chromosome Location | Chr. 10 - 67065896 - 67065981 [+] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000868
miRBase ID: rno-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: Chr. 10 - 67065896 - 67065981 [+] on Build Rnor_6.0 |
Stem-loop ID | abu-mir-193 |
Stem-loop Accession # | MI0033876 |
Stem-loop Sequence | UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042161
miRBase ID: abu-miR-193-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | bta-mir-193a |
Stem-loop Accession # | MI0005014 |
Stem-loop Sequence | UGGGAGCUGAGAGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003795
miRBase ID: bta-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | gga-mir-193a |
Stem-loop Accession # | MI0007564 |
Stem-loop Sequence | AGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGAGGUUCGUGUCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0007740
miRBase ID: gga-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | cgr-mir-193a |
Stem-loop Accession # | MI0033081 |
Stem-loop Sequence | GAGAGCUGGGUCUUUGCGGGCAAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCCUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0041152
miRBase ID: cgr-miR-193a Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | ccr-mir-193a |
Stem-loop Accession # | MI0023355 |
Stem-loop Sequence | AAAGACAUUAAUGUGUUAGAGGUUGGGUCUUUGCGGGCAAGGUGAGUAGUUAAAUUUACUCUCAACUGGCCUACAAAGUCCCAGUUUCUGGCUCAUGUAAUCAUCCUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0026251
miRBase ID: ccr-miR-193a Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | dno-mir-193a |
Stem-loop Accession # | MI0039024 |
Stem-loop Sequence | UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0047722
miRBase ID: dno-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | efu-mir-193a |
Stem-loop Accession # | MI0028845 |
Stem-loop Sequence | AGUUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCUCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0035142
miRBase ID: efu-miR-193a Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | fru-mir-193 |
Stem-loop Accession # | MI0003285 |
Stem-loop Sequence | UGUGUCAGAAGCUGGGUCUUUGCGGGCAAGGUGAGUCCUCAGUUUGUUCAACUGGCCUACAAAGUCCCAGUCUCUGGCUCACGUAACC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002969
miRBase ID: fru-miR-193 Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | cpo-mir-193a |
Stem-loop Accession # | MI0038681 |
Stem-loop Sequence | UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0047071
miRBase ID: cpo-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | eca-mir-193a |
Stem-loop Accession # | MI0012773 |
Stem-loop Sequence | GGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCUUCGGCCCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013026
miRBase ID: eca-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | mze-mir-193 |
Stem-loop Accession # | MI0034175 |
Stem-loop Sequence | UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042391
miRBase ID: mze-miR-193 Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | nbr-mir-193 |
Stem-loop Accession # | MI0034431 |
Stem-loop Sequence | UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042573
miRBase ID: nbr-miR-193 Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | oha-mir-193 |
Stem-loop Accession # | MI0031387 |
Stem-loop Sequence | UGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036780
miRBase ID: oha-miR-193-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | oni-mir-193 |
Stem-loop Accession # | MI0034686 |
Stem-loop Sequence | UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042763
miRBase ID: oni-miR-193 Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | oga-mir-193a |
Stem-loop Accession # | MI0040504 |
Stem-loop Sequence | UGGGUCUUUGCGGGCGAGAUGAGGAUGUCGGAUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049716
miRBase ID: oga-miR-193a Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | ppa-mir-193a |
Stem-loop Accession # | MI0039804 |
Stem-loop Sequence | UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049094
miRBase ID: ppa-miR-193a Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | ptr-mir-193a |
Stem-loop Accession # | MI0008566 |
Stem-loop Sequence | CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUUCUCGGCCCCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0008058
miRBase ID: ptr-miR-193a Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | pha-mir-193a |
Stem-loop Accession # | MI0040283 |
Stem-loop Sequence | UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049518
miRBase ID: pha-miR-193a Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | ssc-mir-193a |
Stem-loop Accession # | MI0013113 |
Stem-loop Sequence | GCGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013895
miRBase ID: ssc-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | ppy-mir-193a |
Stem-loop Accession # | MI0014855 |
Stem-loop Sequence | CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0015788
miRBase ID: ppy-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | pal-mir-193a |
Stem-loop Accession # | MI0032406 |
Stem-loop Sequence | UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGAUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039932
miRBase ID: pal-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | pbv-mir-193a |
Stem-loop Accession # | MI0030265 |
Stem-loop Sequence | GAGCAGCUGGGAGUUUGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUCUUAACUUUGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038990
miRBase ID: pbv-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | ocu-mir-193a |
Stem-loop Accession # | MI0039330 |
Stem-loop Sequence | UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0048304
miRBase ID: ocu-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | mml-mir-193a |
Stem-loop Accession # | MI0007655 |
Stem-loop Sequence | GGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006226
miRBase ID: mml-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | tni-mir-193 |
Stem-loop Accession # | MI0003286 |
Stem-loop Sequence | CAGAAGCUGGGUCUUUGCGGGCAAGGUGAGUCCUCAGUUUGUUCAACUGGCCUACAAAGUCCCAGUGUCUGGCUCACGUGACC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002970
miRBase ID: tni-miR-193 Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | tch-mir-193a-1 |
Stem-loop Accession # | MI0031248 |
Stem-loop Sequence | UGGGUCUUUGCGGGCAAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036584
miRBase ID: tch-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | tch-mir-193a-2 |
Stem-loop Accession # | MI0031268 |
Stem-loop Sequence | UGGGUCUUUGCGGGCAAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036584
miRBase ID: tch-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | dre-mir-193a-1 |
Stem-loop Accession # | MI0002029 |
Stem-loop Sequence | GACAUUAAUGUGUUAGAGGUUGGGUCUUUGCGGGCAAGGUGAGUAGUUAAAUUUACUCUCAACUGGCCUACAAAGUCCCAGUUUCUGGCUCAUGUUAUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0001856
miRBase ID: dre-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |
Stem-loop ID | dre-mir-193a-2 |
Stem-loop Accession # | MI0002030 |
Stem-loop Sequence | GAGAGUGUGUCAGAGGCUGGGUCUUUGCGGGCAAGGUGAGUUUUCCUUUCAUUCAACUGGCCUACAAAGUCCCAGUUUUCGGCCCAUGUGACCAUCUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0001856
miRBase ID: dre-miR-193a-3p Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU Chromosome Location: NA |

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
























miRBase Alias:
Mouse: mmu-miR-193 (v17)
Mouse: mmu-miR-193-3p (v18)
Rat: rno-miR-193-3p (v20)
Rat: rno-miR-193 (v18)
Bos taurus: bta-miR-193a (v13)
Danio rerio: dre-miR-193a (v20)
Gallus gallus: gga-miR-193a (v19)
Mouse: mmu-miR-193-3p (v18)
Rat: rno-miR-193-3p (v20)
Rat: rno-miR-193 (v18)
Bos taurus: bta-miR-193a (v13)
Danio rerio: dre-miR-193a (v20)
Gallus gallus: gga-miR-193a (v19)
