Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | AUAAGACGAACAAAAGGUUUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-208b-3p | MIMAT0004960 | |
| Mouse | mmu-miR-208b-3p | MIMAT0004939 | |
| Bovine | bta-miR-208b | MIMAT0009262 | |
| Callithrix jacchus | cja-miR-208b | MIMAT0039596 | |
| Dasypus novemcinctus | dno-miR-208b-3p | MIMAT0047753 | |
| Dog | cfa-miR-208b | MIMAT0009869 | |
| View More Rows | |||
| Guinea pig | cpo-miR-208b-3p | MIMAT0047108 | |
| Horse | eca-miR-208b | MIMAT0012900 | |
| Monodelphis domestica | mdo-miR-208b-3p | MIMAT0028613 | |
| Pan troglodytes | ptr-miR-208b | MIMAT0008068 | |
| Pig | ssc-miR-208b | MIMAT0013912 | |
| Pongo pygmaeus | ppy-miR-208b | MIMAT0015800 | |
| Rabbit | ocu-miR-208b-3p | MIMAT0048332 | |
| Rhesus monkey | mml-miR-208b-3p | MIMAT0006239 | |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Monodelphis domestica
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-208b (v19)
Mouse: mmu-miR-208b (v17)
Macaca mulatta: mml-miR-208b (v19)
Mouse: mmu-miR-208b (v17)
Macaca mulatta: mml-miR-208b (v19)

Genomic Map