Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | AUAAGACGAACAAAAGGUUUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-208b-3p | MIMAT0004960 |
| Mouse | mmu-miR-208b-3p | MIMAT0004939 |
| Bovine | bta-miR-208b | MIMAT0009262 |
| Callithrix jacchus | cja-miR-208b | MIMAT0039596 |
| Dasypus novemcinctus | dno-miR-208b-3p | MIMAT0047753 |
| Dog | cfa-miR-208b | MIMAT0009869 |
| View More Rows | ||
| Guinea pig | cpo-miR-208b-3p | MIMAT0047108 |
| Horse | eca-miR-208b | MIMAT0012900 |
| Monodelphis domestica | mdo-miR-208b-3p | MIMAT0028613 |
| Pan troglodytes | ptr-miR-208b | MIMAT0008068 |
| Pig | ssc-miR-208b | MIMAT0013912 |
| Pongo pygmaeus | ppy-miR-208b | MIMAT0015800 |
| Rabbit | ocu-miR-208b-3p | MIMAT0048332 |
| Rhesus monkey | mml-miR-208b-3p | MIMAT0006239 |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Monodelphis domestica
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-208b (v19)
Mouse: mmu-miR-208b (v17)
Macaca mulatta: mml-miR-208b (v19)
Mouse: mmu-miR-208b (v17)
Macaca mulatta: mml-miR-208b (v19)

Genomic Map