Assay Details
| Gene Family ID: | MIPF0000033, mir-10 | 
| Mature miRNA Sequence: | ACAGAUUCGAUUCUAGGGGAAU | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-10b-3p | MIMAT0004556 | 
| Alligator mississippiensis | ami-miR-10b-3p | MIMAT0038033 | 
| Cricetulus griseus | cgr-miR-10b-3p | MIMAT0023738 | 
| Eptesicus fuscus | efu-miR-10 | MIMAT0034920 | 
| Goat | chi-miR-10b-3p | MIMAT0035914 | 
| Pteropus alecto | pal-miR-10b-3p | MIMAT0039900 | 
Stem-loop Details
Human
Alligator mississippiensis
Cricetulus griseus
Eptesicus fuscus
Goat
Pteropus alecto
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :