Assay Details
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | AAGACGGGAGGAAAGAAGGGAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-483-5p | MIMAT0004761 |
| Gorilla gorilla | ggo-miR-483 | MIMAT0024191 |
Stem-loop Details
Human
Gorilla gorilla

Genomic Map