Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | CACUAGAUUGUGAGCUGCUGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-28a-3p | MIMAT0004661 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-28* (v17)
Mouse: mmu-miR-28-3p (v18)
Mouse: mmu-miR-28-3p (v18)

Genomic Map