Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAGGACACAUGGUCUACUUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-1197 | MIMAT0005955 | |
| Mouse | mmu-miR-1197-3p | MIMAT0005858 | |
| Bovine | bta-miR-1197 | MIMAT0009966 | |
| Dasypus novemcinctus | dno-miR-1197-3p | MIMAT0048030 | |
| Goat | chi-miR-1197-3p | MIMAT0035917 | |
| Guinea pig | cpo-miR-1197-3p | MIMAT0047404 | |
| View More Rows | |||
| Horse | eca-miR-1197 | MIMAT0013125 | |
| Pan troglodytes | ptr-miR-1197 | MIMAT0007954 | |
| Pongo pygmaeus | ppy-miR-1197 | MIMAT0016149 | |
| Rabbit | ocu-miR-1197-3p | MIMAT0048584 | |
| Sheep | oar-miR-1197-3p | MIMAT0019258 | |
Stem-loop Details
Human
Mouse
Bovine
Dasypus novemcinctus
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Sheep
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Human MicroRNA B Cards v3.0TaqMan® Rodent MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Mouse: mmu-miR-1197 (v17)

Genomic Map