Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAGGACACAUGGUCUACUUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1197 | MIMAT0005955 |
| Mouse | mmu-miR-1197-3p | MIMAT0005858 |
| Bovine | bta-miR-1197 | MIMAT0009966 |
| Dasypus novemcinctus | dno-miR-1197-3p | MIMAT0048030 |
| Goat | chi-miR-1197-3p | MIMAT0035917 |
| Guinea pig | cpo-miR-1197-3p | MIMAT0047404 |
| View More Rows | ||
| Horse | eca-miR-1197 | MIMAT0013125 |
| Pan troglodytes | ptr-miR-1197 | MIMAT0007954 |
| Pongo pygmaeus | ppy-miR-1197 | MIMAT0016149 |
| Rabbit | ocu-miR-1197-3p | MIMAT0048584 |
| Sheep | oar-miR-1197-3p | MIMAT0019258 |
Stem-loop Details
Human
Mouse
Bovine
Dasypus novemcinctus
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Sheep

Genomic Map