Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAAGUAAUUGUGGAUUUUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548n | MIMAT0005916 |
| Callithrix jacchus | cja-miR-548c | MIMAT0039603 |
| Pan troglodytes | ptr-miR-548n | MIMAT0008227 |
Stem-loop Details
Human
Callithrix jacchus
Pan troglodytes

Genomic Map