Assay Details
Mature miRNA Sequence: | UUCACCUCUCUCCAUACUUAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-1016-3p | MIMAT0005029 |
Stem-loop Details
Drosophila melanogaster
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dme-miR-1016-3p |
|---|---|
| miRBase Accession Number: | MI0005877 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCACCUCUCUCCAUACUUAG |
| Species: | Drosophila melanogaster |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUCACCUCUCUCCAUACUUAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-1016-3p | MIMAT0005029 |