Assay Details
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUUUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Anolis carolinensis | aca-miR-499-5p | MIMAT0021980 | |
| Astatotilapia burtoni | abu-miR-499 | MIMAT0042136 | |
| Chrysemys picta | cpi-miR-499-5p | MIMAT0037901 | |
| Cricetulus griseus | cgr-miR-499-5p | MIMAT0023972 | |
| Cyprinus carpio | ccr-miR-499 | MIMAT0026309 | |
| Dasypus novemcinctus | dno-miR-499-5p | MIMAT0047936 | |
| View More Rows | |||
| Gadus morhua | gmo-miR-499-5p | MIMAT0044196 | |
| Guinea pig | cpo-miR-499-5p | MIMAT0047297 | |
| Metriaclima zebra | mze-miR-499 | MIMAT0042375 | |
| Neolamprologus brichardi | nbr-miR-499 | MIMAT0042555 | |
| Oreochromis niloticus | oni-miR-499 | MIMAT0042739 | |
| Pundamilia nyererei | pny-miR-499 | MIMAT0042934 | |
| Python bivittatus | pbv-miR-499-5p | MIMAT0039065 | |
| Rabbit | ocu-miR-499-5p | MIMAT0048489 | |
| Salmo salar | ssa-miR-499b-5p | MIMAT0032661 | |
| Xenopus laevis | xla-miR-499-5p | MIMAT0046607 | |
| Zebrafish | dre-miR-499-5p | MIMAT0003749 | |
Stem-loop Details
Anolis carolinensis
Astatotilapia burtoni
Chrysemys picta
Cricetulus griseus
Cyprinus carpio
Dasypus novemcinctus
Gadus morhua
Guinea pig
Metriaclima zebra
Neolamprologus brichardi
Oreochromis niloticus
Pundamilia nyererei
Python bivittatus
Rabbit
Salmo salar
Xenopus laevis
Zebrafish

Genomic Map