Assay Details
Gene Family ID: | MIPF0000403, MIR482 |
Mature miRNA Sequence: | UUUUUCCUACUCCGCCCAUACC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR472-3p | MIMAT0003931 |
| Arabidopsis lyrata | aly-miR472-3p | MIMAT0017590 |
Stem-loop Details
Arabidopsis
Arabidopsis lyrata
More Information
Related Products
miRBase Alias:
Arabidopsis thaliana: ath-miR472 (v20)
Arabidopsis lyrata: aly-miR472 (v18)
Arabidopsis lyrata: aly-miR472 (v18)

Genomic Map