Assay Details
Mature miRNA Sequence: | UGGCAGUGUUUCUCCCCCAACUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-1824-5p | MIMAT0006591 |
Stem-loop Details
Caenorhabditis elegans
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cel-miR-1824-5p |
|---|---|
| miRBase Accession Number: | MI0007986 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGCAGUGUUUCUCCCCCAACUU |
| Species: | Caenorhabditis elegans |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UGGCAGUGUUUCUCCCCCAACUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-1824-5p | MIMAT0006591 |