Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGUAGUGAUGUUUAG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-499-5p | MIMAT0003367 |
Stem-loop Details
Chicken

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | gga-miR-499 |
---|---|
miRBase Accession Number: | MI0003710 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUAAGACUUGUAGUGAUGUUUAG |
Species: | Chicken |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGUAGUGAUGUUUAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-499-5p | MIMAT0003367 |