More Information
Updated Mapping Information

miRBase Alias:
Gallus gallus: gga-miR-1566 (v20)

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | gga-miR-1566 |
---|---|
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CGGGACAGGACGGCUCUCGGA |
Product Type: | TaqMan™ MicroRNA Assay |