Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUCAUUCACGGACAACACUUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dog | cfa-miR-382 | MIMAT0006724 |
Eptesicus fuscus | efu-miR-382 | MIMAT0035040 |
Pteropus alecto | pal-miR-382-3p | MIMAT0039916 |
Stem-loop Details
Dog
Eptesicus fuscus
Pteropus alecto
