Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAAUAUGGUCCACGUCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dasypus novemcinctus | dno-miR-380-3p | MIMAT0047863 |
Dog | cfa-miR-380 | MIMAT0006717 |
Guinea pig | cpo-miR-380-3p | MIMAT0047224 |
Pteropus alecto | pal-miR-380-3p | MIMAT0040048 |
Stem-loop Details
Dasypus novemcinctus
Dog
Guinea pig
Pteropus alecto
