Assay Details
Mature miRNA Sequence: | ACCUCUCCAUCAUGCAUGACA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Amphimedon queenslandica | aqu-miR-2015-3p | MIMAT0009689 |
Stem-loop Details
Amphimedon queenslandica

More Information
Related Products
miRBase Alias:
Amphimedon queenslandica: aqu-miR-2015 (v18)
