Assay Details
Gene Family ID: | MIPF0000313, mir-306 |
Mature miRNA Sequence: | GAGGGUCGCUCUGUGCCUGUGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila mojavensis | dmo-miR-306-3p | MIMAT0008649 |
Drosophila virilis | dvi-miR-306-3p | MIMAT0008914 |
Stem-loop Details
Drosophila mojavensis
Drosophila virilis

More Information
Related Products
miRBase Alias:
Drosophila mojavensis: dmo-miR-306* (v18)
Drosophila virilis: dvi-miR-306* (v18)
Drosophila virilis: dvi-miR-306* (v18)
