Assay Details
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | UGAGAUUCAACUCCUCCAACUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Capitella teleta | cte-miR-1175-3p | MIMAT0009550 |
| Lottia gigantea | lgi-miR-1175-3p | MIMAT0009603 |
| Melibe leonina | mle-miR-1175-3p | MIMAT0048798 |
Stem-loop Details
Capitella teleta
Lottia gigantea
Melibe leonina
More Information
Related Products
miRBase Alias:
Comamonas testosteroni: cte-miR-1175 (v18)
Lachnoanaerobaculum gingivalis: lgi-miR-1175 (v18)
Lachnoanaerobaculum gingivalis: lgi-miR-1175 (v18)

Genomic Map