Assay Details
Mature miRNA Sequence: | CCUCUUUACAUUGUUGUGCUGUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Nematostella vectensis | nve-miR-2030-3p | MIMAT0009781 |
Stem-loop Details
Nematostella vectensis
More Information
Related Products
miRBase Alias:
Nematostella vectensis: nve-miR-2030* (v18)

Genomic Map