Assay Details
Mature miRNA Sequence: | UACCGGAGUCUUUUCUGUG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Nematostella vectensis | nve-miR-2051-3p | MIMAT0009829 |
Stem-loop Details
Nematostella vectensis

More Information
Related Products
miRBase Alias:
Nematostella vectensis: nve-miR-2051 (v18)
