Assay Details
Gene Family ID: | MIPF0000102, MIR408 |
Mature miRNA Sequence: | CUGCACUGCCUCUUCCCUGGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Brachypodium distachyon | bdi-miR408-3p | MIMAT0027039 | |
Carica papaya | cpa-miR408 | MIMAT0031823 | |
Digitalis purpurea | dpr-miR408 | MIMAT0023538 | |
Rice | osa-miR408-3p | MIMAT0001079 | |
Ricinus communis | rco-miR408 | MIMAT0014204 | |
Saccharum officinarum | sof-miR408a | MIMAT0001667 | |
View More Rows | |||
Saccharum officinarum | sof-miR408b | MIMAT0001668 | |
Saccharum officinarum | sof-miR408c | MIMAT0001669 | |
Saccharum officinarum | sof-miR408d | MIMAT0001670 | |
Saccharum sp. | ssp-miR408a | MIMAT0020297 | |
Saccharum sp. | ssp-miR408d | MIMAT0020296 | |
Sorghum bicolor | sbi-miR408 | MIMAT0011361 | |
Triticum aestivum | tae-miR408 | MIMAT0005350 | |
Zea mays | zma-miR408a | MIMAT0001748 | |
Zea mays | zma-miR408b-3p | MIMAT0014026 |
Stem-loop Details
Brachypodium distachyon
Carica papaya
Digitalis purpurea
Rice
Ricinus communis
Saccharum officinarum
Saccharum sp.
Sorghum bicolor
Triticum aestivum
Zea mays

More Information
Related Products
miRBase Alias:
Oryza sativa: osa-miR408 (v18)
Zea mays: zma-miR408b (v18)
Zea mays: zma-miR408 (v18)
Zea mays: zma-miR408b (v18)
Zea mays: zma-miR408 (v18)
