Assay Details
Mature miRNA Sequence: | AACUGAUUUUGUUUGGGA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ornithorhynchus anatinus | oan-miR-1358 | MIMAT0006990 |
Stem-loop Details
Ornithorhynchus anatinus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | oan-miR-1358 |
---|---|
miRBase Accession Number: | MI0006776 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AACUGAUUUUGUUUGGGA |
Species: | Ornithorhynchus anatinus |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | AACUGAUUUUGUUUGGGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ornithorhynchus anatinus | oan-miR-1358 | MIMAT0006990 |