Assay Details
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | AUGACCUAUGAAUUGACAGACA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-215 | MIMAT0003797 |
| Callithrix jacchus | cja-miR-215 | MIMAT0039385 |
| Cricetulus griseus | cgr-miR-215-5p | MIMAT0023853 |
| Dasypus novemcinctus | dno-miR-215-5p | MIMAT0047760 |
| Monodelphis domestica | mdo-miR-215 | MIMAT0012751 |
Stem-loop Details
Bovine
Callithrix jacchus
Cricetulus griseus
Dasypus novemcinctus
Monodelphis domestica

Genomic Map